Learn More
MSC CHEMWORLD All in One Boiler Chemical, Hard Water, 55 Gallons
Catalog No. 502837986
Encompass
This all-in-one all-polymer internal treatment product is designed for boiler systems with hard water conditions and unsoftened make-up water. It simplifies chemical feeding and testing, including polymer for deposit control, alkalinity building, and sulfite for oxygen removal. It also helps remove existing boiler scale.
  • All-in-one all-polymer internal treatment product.
  • Includes polymer for deposit control.
  • Contains alkalinity builder for maintaining boiler alkalinity.
  • Includes sulfite for removing oxygen from boiler water.
  • Designed for easy chemical feeding and testing.
  • Ideal for gradual removal of existing boiler scale.
  • Protects metal in the steam boiler system against corrosion.
  • Reduces the risk of oxygen corrosion by maintaining sulfite residual.
  • Prevents corrosion and scale formation.
  • Simplifies boiler chemical treatment.
Catalog No. 50-283-7986 Supplier MSC Supplier No. 82931684
This all-in-one all-polymer internal treatment product is designed for boiler systems with hard water conditions and unsoftened make-up water. It simplifies chemical feeding and testing, including polymer for deposit control, alkalinity building, and sulfite for oxygen removal. It also helps remove existing boiler scale.
  • All-in-one all-polymer internal treatment product.
  • Includes polymer for deposit control.
  • Contains alkalinity builder for maintaining boiler alkalinity.
  • Includes sulfite for removing oxygen from boiler water.
  • Designed for easy chemical feeding and testing.
  • Ideal for gradual removal of existing boiler scale.
  • Protects metal in the steam boiler system against corrosion.
  • Reduces the risk of oxygen corrosion by maintaining sulfite residual.
  • Prevents corrosion and scale formation.
  • Simplifies boiler chemical treatment.