Learn More
Electron Microscopy Sciences Hardened Steel Deburring Tool Set
SDP
Catalog No. 5026446
Encompass
Made from hardened steel, this tool has three razor sharp cutting edges ranging from a very sharp point to one that is approximately 10 mm in length. These nickel plated triangular tools are mounted in red and black plastic handles. Tool length is 30 mm by 2.1 mm across (1.2"x.08"). The overall length of the tool is 154 mm (5.7").2 tools/set.
Catalog No. 50-264-46 Supplier Electron Microscopy Sciences Supplier No. 62161SD
Made from hardened steel, this tool has three razor sharp cutting edges ranging from a very sharp point to one that is approximately 10 mm in length. These nickel plated triangular tools are mounted in red and black plastic handles. Tool length is 30 mm by 2.1 mm across (1.2"x.08"). The overall length of the tool is 154 mm (5.7").2 tools/set.