Learn More
Enterprise Technology Solutions SecureFit H-Series Hard Hats, H-800 Hat with UV Indicator, 4-Point Pressure Diffusion Ratchet Suspension, White
SDP
Catalog No. 502519447
Encompass_Preferred
SecureFit H-Series Hard Hats, H-800 Hat with UV Indicator, 4-Point Pressure Diffusion Ratchet Suspension, White
Catalog No. 50-251-9447 Supplier Enterprise Technology Solutions Supplier No. MMMH801SFRUV
SecureFit H-Series Hard Hats, H-800 Hat with UV Indicator, 4-Point Pressure Diffusion Ratchet Suspension, White