Learn More
Hardigg™ iM2050 Storm Cases
Catalog No. 191305094
Change view
Click to view available options
Product Type:
Cubed Storm Case
Empty Storm Case
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-130-5094 Cubed Storm Case
19-130-5083 Empty Storm Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-130-5094 Supplier Hardigg™ Supplier No. IM205040001
Specifications
Product Type Cubed Storm Case
Color Orange
Material HPX High Performance Resin
Closure Type 2 press and pull latches, 2 padlockable hasps, flush hinges
Includes Double-layered, soft-grip handle, vortex valve
Length (English) Interior 9.5 in.
Length (Metric) Interior 11.9 cm
Width (English) Interior 7.5 in.
Width (Metric) Interior 19.05 cm