Learn More
Bullard™ AboveView™ HDPE Full Brim Hard Hat with Flex-Gear™ 4 Point Ratchet Suspension
SDP
Catalog No. 19160652 Shop All Bullard Company Products
Change view
Click to view available options
Cobalt plated black hardware; Black
Catalog No.
19-160-652 Cobalt plated black hardware; Black
1 options
This item is not returnable. View return policy
Catalog No. 19-160-652 Supplier Bullard™ Supplier No. AVRDRC
This item is not returnable. View return policy
Clear See-Through Visor And Cotton Brow Pad
Featuring a see-through visor that allows workers to see hazards overhead, the patented AboveView Full-Brim Hard Hat is designed with a wide brim for increased protection from the elements and comes equipped with accessory slots for a variety of attachments. ANSI/ISEA Classification: Z89.1, Type I, Class E and G. AboveView comes with all the standard Bullard hard hat comfort features including, 3/4 in. seamless suspension, large absorbent cotton or easy-to-clean vinyl brow pad, Flex-Gear ratchet suspension, and front, side, and rear vertical height adjustment settings.
  • Replaceable See-Through Visor
  • Vertical Adjustments (two front and two rear)
  • Nylon Crown Straps
  • Large, replaceable, absorbent Cotton or easy-to-clean Vinyl brow pad
  • Cushioned Nape
  • Large, easy-to-turn Flex-Gear™ ratchet knob
  • Accessory Slots
Specifications
Color Red
Material HDPE
Product Type Hard Hat