Learn More
Sanford Distributing Co HARDMAN EPOXY/CARTON OF 100
Catalog No. NC9541353
Encompass_Preferred
EPOXY Red ,Extra-fast setting for quick repairs,Excellent for bonding wood,glass,metal,stone,concrete, ceramics and leather,Work time: 3 minutes,Handling strength: 15-30 minutes,Color: Light Amber,Carton of 100
Catalog No. NC9541353 Supplier Sanford Distributing Co Supplier No. 04001
EPOXY Red ,Extra-fast setting for quick repairs,Excellent for bonding wood,glass,metal,stone,concrete, ceramics and leather,Work time: 3 minutes,Handling strength: 15-30 minutes,Color: Light Amber,Carton of 100