Learn More
Hach Company Hardness 2 Indicator Solution, 100 mL MDB
Supplier:  Hach Company 42532
Encompass
ManVer (Calmagite) Hardness Indicator, for hardness and chelant determination by titration. 100 mL Marked Dropping Bottle
Catalog No. 50-199-4284