Learn More
Hach Company Hardness 3 Solution, 1 L
Supplier:  Hach Company 42653
Encompass
EDTA titrant solution for hardness determination by drop-count titration. 0.03 N (0.015 M) 1 liter bottle
Catalog No. 50-199-4289