Learn More
Hach Company Hardness 3 Solution, 100 mL MDB
Supplier:  Hach Company 42632
Encompass
EDTA titrant solution for hardness determination by drop-count titration. 0.03 N (0.015 M) 100 mL Marked Dropping Bottle
Catalog No. 50-199-4287