Learn More
LaMarKa Chemical Company Hardness Buffer (Magnesium Free) contains Ammonium Hydroxide & Ammonium Chloride, 4l
Catalog No. 504400822
Encompass_Preferred
Hardness Buffer (Magnesium Free) contains Ammonium Hydroxide & Ammonium Chloride, 4l Plastic Jug
Catalog No. 50-440-0822 Supplier LaMarKa Chemical Company Supplier No. 00006585167532436
Hardness Buffer (Magnesium Free) contains Ammonium Hydroxide & Ammonium Chloride, 4l Plastic Jug