Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardness buffer 1, pH 10.1±0.1 at 20°, Lovibond™

Catalog No. 09534718 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
65 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-534-718 65 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-534-718 Supplier Tintometer Group Supplier No. R50042
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Buffer Solution Hardness 1, pH 10.1±0.1 at 20°
CAS 64-19-7, 7732-18-5, 124-68-5
Color Yellow
Physical Form Liquid
Quantity 65 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.