Learn More
Lovibond™ Hardness Ca Mg MR Tube Test
Catalog No. 01670227 Shop All Tintometer Group Products
Change view
Click to view available options
No. of Tests:
100
Catalog No. No. of Tests
01-670-227 100
1 options
This item is not returnable. View return policy
Catalog No. 01-670-227 Supplier Lovibond™ Supplier No. 2423960
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ideal for analyzing feedwater of cooling systems, this reagent set is used to determine the concentration of calcium and magnesium hardness in a water sample with a single method.
  • This easy to use colorimetric test contains 4 liquid reagents for performing up to 26 tests.
Specifications
Analytes Calcium and Magnesium Hardness
For Use With (Application) Pre-Treatment of RO Water and Softening Systems
For Use With (Equipment) Lovibond Spectrophotometers and Colorimeters
Includes CA MG Hardness Solution 1, CA MG Hardness Solution 2, CA MG Hardness Solution 3 and CA MG Hardness Solution 4
No. of Tests 100
Range 0.05 to 4 mg/L CaCO3
Test Method Calmagite
Type Colorimetric Test