Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QualityCheck Hardness Sample, Inorganic, Quantitative, 500 mL

Catalog No. USQCI720
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-QCI-720 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-QCI-720 Supplier Agilent Technologies Supplier No. QCI720
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Hardness Drinking Water/Wastewater Standard
Packaging Poly Bottle
Name Note ULTRAcheck Blind QC Standard
Traceability to NIST Yes
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Component concentrations in ready-to-use solution:
Calcium    •10-150mg/L
Magnesium    •5-50mg/L
Hardness (as CaCO3)    •45-575mg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.