Learn More
Taylor Chemical HARDNESS IND PWD 1/4LB
Catalog No. NC9129153
Encompass_Preferred
This item has a minimum qty of 4 per supplier requirements.
Hardness Indicator Powder, Family: Organic dye or sucrose mixture, Specific Gravity: 1.6, Irritant, Appearance: Purple crystals, Sucrose CAS No.: 57-50-1 > 90%, Eriochrome black T CAS No.: 1787-61-7 <1%, 0.25 lb., Volatile by volume: 1%
Catalog No. NC9129153 Supplier Taylor Chemical Supplier No. R0620J
This item has a minimum qty of 4 per supplier requirements.
Hardness Indicator Powder, Family: Organic dye or sucrose mixture, Specific Gravity: 1.6, Irritant, Appearance: Purple crystals, Sucrose CAS No.: 57-50-1 > 90%, Eriochrome black T CAS No.: 1787-61-7 <1%, 0.25 lb., Volatile by volume: 1%