Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardness Standard 250mg/L, Reagents

Catalog No. 01335156
Change view
Click to view available options
Quantity:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-156 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-156 Supplier Reagents Holdings Llc Supplier No. CH00090010F
Add to Cart
Add to Cart

Hardness Standard 250mg/L, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Hardness Standard 250 mg/L
Packaging Cubitainer
CAS 7732-18-5, 10035-04-8
Specific Gravity 1.0004
Physical Form Liquid
Color Colorless
pH 4.5 to 8.5
Boiling Point 100°C
Solubility Completely miscible
Quantity 10 L
Percent Purity 0.025%
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.