Learn More
HANNA INSTRUMENTS INC Total Hardness Medium Range Colorimetric Reagents (100 tests)
SDP
Catalog No. NC1710700
Encompass
The HI93735-01 are reagents for the medium range colorimetric determination of total hardness. There are enough reagents for 100 tests to be used with Hanna's HI96735 Total Hardness EPA Portable Photometer. These high quality reagents are manufactured in our state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet for traceability.Pre-made reagents for ease of usePrepared with high purity chemicalsMarked with expiration date and lot number for traceability
Catalog No. NC1710700 Supplier HANNA INSTRUMENTS INC Supplier No. HI9373501
The HI93735-01 are reagents for the medium range colorimetric determination of total hardness. There are enough reagents for 100 tests to be used with Hanna's HI96735 Total Hardness EPA Portable Photometer. These high quality reagents are manufactured in our state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet for traceability.Pre-made reagents for ease of usePrepared with high purity chemicalsMarked with expiration date and lot number for traceability