Learn More
Bio Rad Laboratories HARDSHELL 96WELL PCR PLTS 50PK
Catalog No. NC1711052
Encompass
Pkg of 50, white shell/white well PCR plate for higher fluorescent signals, rigid 2-component design
Catalog No. NC1711052 Supplier Bio Rad Laboratories Supplier No. HSP9655
Pkg of 50, white shell/white well PCR plate for higher fluorescent signals, rigid 2-component design