Learn More
Medicus Health Hardside Cooler, Small 13 Qt
SDP
Catalog No. NC2171640
Encompass
∙ The hard side Blood Transport Cooler is perfect for transporting specimens to the lab and maintaining temperatures ∙ Great option for Blood Banks to send units to the ER or Surgery to help prevent units from returning over temperature∙ Hard side blood transport coolers have feet and can be stacked for storage∙ Stainless Steel hardware on this small hard side cooler for medical transport will provide many years of durable service∙ Convenient handle on top and heavy duty strap can be used to secure lid during transport or be used as a shoulder strap∙ Constructed of durable polypropylene for all types of specimen transport needs∙ Non-absorbent surface easy to clean and disinfect∙ Airtight closure seal is provided by an EVA Gasket Seal recessed into the lid∙ Internal Dimensions: 12"L x 7"W x 8.5"H; Ext Dimensions: 15"L x 10"W x 11"H
This item is not returnable. View return policy
Catalog No. NC2171640 Supplier Medicus Health Supplier No. 5318M1
∙ The hard side Blood Transport Cooler is perfect for transporting specimens to the lab and maintaining temperatures ∙ Great option for Blood Banks to send units to the ER or Surgery to help prevent units from returning over temperature∙ Hard side blood transport coolers have feet and can be stacked for storage∙ Stainless Steel hardware on this small hard side cooler for medical transport will provide many years of durable service∙ Convenient handle on top and heavy duty strap can be used to secure lid during transport or be used as a shoulder strap∙ Constructed of durable polypropylene for all types of specimen transport needs∙ Non-absorbent surface easy to clean and disinfect∙ Airtight closure seal is provided by an EVA Gasket Seal recessed into the lid∙ Internal Dimensions: 12"L x 7"W x 8.5"H; Ext Dimensions: 15"L x 10"W x 11"H