Learn More
Medchemexpress LLC Hardwickiic acid | 1782-65-6 | 1 MG
SDP
Catalog No. 5000418325
Encompass_Preferred
Hardwickiic acid is a diterpenoid compound that can block tetrodotoxin-sensitive voltage-dependent sodium channels. It possesses multiple activities including insecticidal, antinociceptive, and neurotransmitter release-regulating effects. It is used in the study of neurological diseases.
  • Neuroprotectant
  • Blocks tetrodotoxin-sensitive voltage-dependent sodium channels
  • Possesses insecticidal activity
  • Exhibits antinociceptive activity
  • Regulates neurotransmitter release
Catalog No. 50-004-18325 Supplier Medchemexpress LLC Supplier No. HYN39901MG
Hardwickiic acid is a diterpenoid compound that can block tetrodotoxin-sensitive voltage-dependent sodium channels. It possesses multiple activities including insecticidal, antinociceptive, and neurotransmitter release-regulating effects. It is used in the study of neurological diseases.
  • Neuroprotectant
  • Blocks tetrodotoxin-sensitive voltage-dependent sodium channels
  • Possesses insecticidal activity
  • Exhibits antinociceptive activity
  • Regulates neurotransmitter release