Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Weiman Floor Cleaner, Finished Wood, 32 fl. oz., Squeeze Bottle

Catalog No. 502835934
Encompass

This floor cleaner is specifically formulated for finished wood surfaces. Provided in a 32 fl. oz. squeeze bottle, it offers a convenient and effective solution for routine maintenance. Its pH-balanced and solvent-based composition ensures a thorough cleaning without harming the floor's finish.

  • Product type: Floors
  • Container type: Squeeze Bottle
  • Container size: 32 fl. oz.
  • Material application: Finished Wood
  • Composition: Solvent Based
  • Formula type: pH Balanced

Catalog No. 50-283-5934 Supplier MSC Supplier No. 74039082
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This floor cleaner is specifically formulated for finished wood surfaces. Provided in a 32 fl. oz. squeeze bottle, it offers a convenient and effective solution for routine maintenance. Its pH-balanced and solvent-based composition ensures a thorough cleaning without harming the floor's finish.

  • Product type: Floors
  • Container type: Squeeze Bottle
  • Container size: 32 fl. oz.
  • Material application: Finished Wood
  • Composition: Solvent Based
  • Formula type: pH Balanced

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.