Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Bovine Blood Agar
SDP

Catalog No. 02686210 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Quantity:
17 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-686-210 17 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-686-210 Supplier Hardy Diagnostics™ Supplier No. A188
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For detection of organisms that may cause mastitis in dairy cattle, such as Staphylococcus aureus and Streptococcus agalactiae

  • Bovine Blood Agar: 5%
  • Bovine mastitis determination
  • Staphylococcus determination
  • 15 x 100mm plate
  • Short shelf life: 100 days

Specifications

Content And Storage 2°C to 8°C
Form Solid
Product Type Blood Agar
Packaging Type Plate
Quantity 17 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.