Learn More
Description
Includes
Z140A – Strep B Carrot Broth, 13 x 100mm Tube, 4mL (20 tubes); Z140B – Strep B Carrot Broth Tiles, 1/4 in. x 1/2 in. (20 tiles/jar)
For processing clinical specimens for culture of group B Streptococcus (GBS).
- Increased sensitivity
- Decreased turn around time
- Overall lower cost
- Results as early as six hours after inoculation
- See Z140T if extra tiles are needed
US Patent No. 8,313,938 B1 and US Patent No. 8,518,688 B1
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | beta-haemolytic Streptococci (Group B) |
| Sample Type | Clinical Specimens |
| Type | Strep B Carrot Broth Kit |
| Includes | Strep B Carrot Broth Tiles, 1/4 in. x 1/2 in. (20 tiles/jar) |
| Quantity | 20 Tests |
| Test Time | 360 min. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
