Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ HardyCHROM™ Candida and Auris Chromogenic Medium
SDP

Catalog No. 23001752 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-752 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-752 Supplier Hardy Diagnostics™ Supplier No. G343
Add to Cart
Add to Cart
This item is not returnable. View return policy

Candida + auris is recommended for the selective isolation and differential indentification of Candida species

  • The medium allows for the differentiation of Candida albicans, Candida tropicalis, Candida krusei, and Candida auris based on colony morphology, color, and positive UV fluorescence.
  • Candida auris produces white colonies with teal to teal-green 'bullseye' centers that are positive for UV fluorescence for easy differentiation from other Candida species.
  • Distinct color change read-out
  • Most strains will exhibit colors within 48 hours
  • Colonies of Candida glabrata can be subbed directly to Rapid Trehalose Fermentation Broth or GlabrataQuick for confirmation

Specifications

Appearance Opaque
Color White
Content And Storage 2°C to 8°C
Country of Origin United States
Additive Chloramphenicol, Chromogenic Mixture, Titanium Dioxide
Detects Candida albicans, Candida tropicalis, Candida krusei, Candida auris, Candida glabrata
Form Prepared Medium
Format Plate
Manufacturing Origin United States
Product Type Agar Plate
Target Organism Class Yeast
Test Type Microbial Differentiation
Fill Volume 18 mL
Formulation Glucose - 20.0gm, Peptone - 10.0gm, Chromogenic Mixture - 2.15gm, Titanium Dioxide - 2.0gm, Chloramphenicol - 0.5gm, Agar - 15.0gm
For Use With (Application) Selective Isolation, Differential Identification
Incubation Conditions 35°C
Incubation Time 24 to 48 hr.
Material Polystyrene
No. per Pack 10
Packaging Type Plates, Bag
Quantity 10 Ea.
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Depth (Metric) 15 mm
Diameter (Metric) 100 mm
Species Candida species, Candida alibicans
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.