Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ HardyCHROM™ CRE Chromogenic Medium
SDP

Catalog No. 23001781 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Product Type: Agar Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-001-781 Product Type: Agar Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-781 Supplier Hardy Diagnostics™ Supplier No. G323
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CRE is a selective chromogenic agar medium intended for the detection from stool specimens of E. coli that are non-susceptible to carbapenems as pink colonies and KES that are non-susceptible to carbapenems as blue colonies.

  • HardyCHROM™ CRE is intended as an aid in the detection, identification of colonization, and control of these bacteria in a healthcare setting
  • Results can be interpreted after incubation for 18-24 hours
  • Subculture to non-selective medium is required for confirming identification, antimicrobial susceptibility testing, and epidemiological typing
  • Non-susceptible E. coli produces colonies that are rose to magenta in color with darker pink centers
  • Klebsiella, Enterobacter,and Serratia spp.produce large, dark blue colonies (with or without a pink halo), are presumptive positive for carbapenem non-susceptible KES (Klebsiella aerogenes, Klebsiella oxytoca, Klebsiella pneumoniae, Enterobacter cloacaecomplex, and Serratia marcescens)
  • Colonies that are not pink to magenta, blue, or blue with pink halos are negative - No carbapenem non-susceptible Escherchia coli or KES detected
  • This media is to be stored and incubated in the dark

Specifications

Appearance Opaque
Color White
Content And Storage 2°C to 8°C
Additive Chromogenic Mixture, Bile Salts, Selective Agents
Detects Carbapenem non-susceptible Escherichia coli, Klebsiella aerogenes, Klebsiella oxytoca, Klebsiella pneumoniae, Enterobacter cloacae complex, Serratia marcescens
Form Agar
Format Plate
Product Type Agar Plate
Target Organism Class Gram-negative Bacteria
Test Type Carbapenem-resistant Enterobacterales (CRE)
Fill Volume 18 mL
Formulation Peptones - 20.0gm, Sodium Chloride - 5.0gm, Chromogenic Mixture - 5.0gm, Bile Salts - 1.5gm, Selective Agents - 1.0gm, Agar - 13.5gm
For Use With (Application) Detection, Identification of colonization
Incubation Conditions 35°C
Incubation Time 18 to 24 hr.
Quantity 10 x 18 mL
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Depth (Metric) 15 mm
Diameter (Metric) 100 mm
Species Klebsiella species
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.