Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ HardyCHROM™ CRE/ESBL, Chromogenic Medium, BiPlate
SDP

Catalog No. 21200327 Shop All Hardy Diagnostics Products
Change view
Click to view available options
CRE/ESBL Chromogenic Agar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
21-200-327 CRE/ESBL Chromogenic Agar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-200-327 Supplier Hardy Diagnostics™ Supplier No. J28
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective and differential chromogenic agar medium intended for the qualitative and presumptive detection from stool specimens of E. coli.

  • HardyCHROM™ CRE is a selective and differential chromogenic agar medium intended for the qualitative and presumptive detection from stool specimens of Escherichia coli that are non-susceptible to carbapenems as pink colonies and KES (Klebsiella aerogenes, Klebsiella oxytoca, Klebsiella pneumoniae, Enterobacter cloacae complex, and Serratia marcescens) that are non-susceptible to carbapenems as blue colonies.
  • HardyCHROM™ ESBL is a selective and differential chromogenic medium that is intended for the qualitative and presumptive detection from stool specimens of Enterobacterales that are potentially non-susceptible to ceftazidime and cefpodoxime, and ESBL-producing Escherichia coli, Klebsiella pneumoniae, and Klebsiella oxytoca.
  • Features and Benefits:
    • Optically clear polystyrene for distortion-free viewing.
    • Heavy-duty to reduce breakage during shipping.
    • Stacking rings: Polystyrene Petri plates feature raised rings for stacking ability and to prevent sliding.
    • Breathable packaging: Packaged in sleeves of 10 plates in a breathable bag that prevents buildup of condensation and excess moisture.
  • This media is to be stored and incubated in the dark.

Specifications

Country of Origin United States
Detects Carbapenem Resistant Enterobacterales, Extended-spectrum beta-lactamase (ESBL)-producing Enterobacterales
Format BiPlate
Manufacturing Origin United States
Media Type Chromogenic Agar Medium
Product Type Agar Plate
Sterility Non-sterile
Target Organism Class Multidrug-resistant, Gram-negative Organisms
Test Type Microbial Detection
For Use With (Application) Microbial Detection
Material Polystyrene
Packaging Type Bag, Sleeve
Quantity 10 Plates
Dimensions (Dia. x H) 100 x 15 mm
Species Escherichia coli
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.