Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ HardyCHROM™ ECC Chromogenic Medium
SDP

Catalog No. 23001820 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Product Type: Contact Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-001-820 Product Type: Contact Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-820 Supplier Hardy Diagnostics™ Supplier No. G303
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

HardyCHROM™ ECC is a chromogenic media recommended for the detection, differentiation, and enumeration of Escherichia coli and other coliforms in food, water, or enviromental samples based on colony color.

  • Routine testing to assess the sanitary quality of food and water is directed at the detection and enumeration of indicator organisms rather than pathogens. The coliform group of organisms is recognized as the principal indicator of unsanitary conditions.
  • E. coli can be identified as pink to violet colored colonies on the plate
  • Other coliform bacteria will appear as turquoise colonies
  • Organisms other than coliforms or E. coli (including approximately 4% of E. coli strains and most O157 strains) will appear as white or colorless colonies
  • Selective agents have been added to inhibit the growth of gram-positive and other non-coliform bacteria
  • Each contact plate has a specified grid molded into the bottom of the plate for enumeration of microbial colonies growing on a variety of surfaces.
  • This media is to be stored and incubated in the dark

Specifications

Appearance Slightly Opalescent
Color Light Amber
Content And Storage 2°C to 8°C
Country of Origin United States
Detects E. coli, Coliforms
Form Prepared Medium
Format Plate
Manufacturing Origin US with Globally Sourced Materials
Media Type Chromogenic agar medium
Product Type Contact Plate
Target Organism Class Coliforms, E. coli, Gram-negative Rods
Test Type Environmental Test
Fill Volume 18 mL
Formulation Peptones - 8.0g, Sodium Chloride - 5.0g, Chromogenic Mixture - 5.0g, Selective Agents - 6.0g, Agar - 15.0g
For Use With (Application) Food, Water, Environmental Samples
Incubation Conditions 35°C
Incubation Time 18 to 24 hr.
Packaging Type Bag
Quantity 10 Plates
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Klebsiella pneumoniae subsp. pneumoniae, Escherichia coli, Enterobacter cloacae subsp. cloacae, Enterococcus faecalis
Protein Source Peptones
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.