Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ HardyCHROM™ ESBL Chromogenic Medium
SDP

Catalog No. 23001745 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-745 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-745 Supplier Hardy Diagnostics™ Supplier No. G321
Add to Cart
Add to Cart
This item is not returnable. View return policy

ESBL is a selective chromogenic medium that is intended for the detection from stool specimens of Enterobacterales that are potentially non-susceptible to ceftazidime and cefpodoxime and Extended-spectrum beta-lactamase (ESBL)-producing E. coli, Klebsiella pneumoniae , and Klebsiella oxytoca.

  • ESBL or Extended-Spectrum Beta-Lactamases refer, to a group of enzymes produced by some species of bacteria that can mediate the organism's resistance to one or more broad spectrum penicillins and cephalosporin antibiotics
  • This breakthrough culture media will help screen for ESBL producers quickly and more efficiently from stool specimens
  • Distinct color change read-out
  • Bright color development
  • Results in as little as 18 hours

Specifications

Appearance Translucent
Color Light Amber
Detects Enterobacterales, ESBL-producing Escherichia coli, Klebsiella pneumoniae, Klebsiella oxytoca, Proteus mirabilis
Form Prepared Medium
Format Plate
Media Type Chromogenic Medium
Product Type Agar Plate
Target Organism Class Gram-negative Bacteria
Test Type Microbial Detection
Fill Volume 18 mL
For Use With (Application) Screening for ESBL-producing organisms
Incubation Conditions 35°C
Incubation Time 24 hr.
No. per Pack 10
Packaging Type Pack
Quantity 10 Ea.
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Klebsiella species, Escherichia coli, Proteus species
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.