Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ HardyCHROM™ Group A Strep
SDP

Catalog No. 23001789 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Product Type: Agar Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-001-789 Product Type: Agar Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-789 Supplier Hardy Diagnostics™ Supplier No. G337
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

HardyCHROM Group A Strep Agar is a chromogenic medium recommended for the selective cultivation and differential isolation of Group A Streptococcus from specimens based on colony color

  • Group A Strep Agar contains organic nitrogen, particularly amino acids and long-chain polypeptides, from peptones and yeast extract. These, along with additional growth factors, render the medium highly nutritious and help to counteract selective agents added to inhibit unwanted flora
  • Optically clear, heavy-duty polystyrene mono plates for distortion-free viewing and reduced breakage during shipping
  • Red colonies on a white background make detection easy
  • Highly Selective

Specifications

Appearance Opaque
Color White to Off-white
Content And Storage 2°C to 8°C
Additive Growth Factors, Selective Agents, Chromogenic Mixture
Detects Group A Streptococcus (S. pyogenes)
Form Prepared Medium
Format Plate
Media Type Agar
Product Type Agar Plate
Target Organism Class Gram-positive Organisms
Test Type Microbial Detection
Fill Volume 18 mL
Formulation Peptones - 18.0g, Salts - 8.0g, Yeast Extract - 4.0g, Growth Factors - 3.0g, Carbohydrate - 3.0g, Selective Agents - 2.2g, Chromogenic Mixture - 1.7g, Agar - 13.0g
For Use With (Application) Selective Cultivation and Differential Isolation
For Use With (Equipment) CO2 Incubator, Sealed Jar
Incubation Conditions 35°C
Incubation Time 24 hr.
Material Polystyrene
No. per Pack 10
Packaging Type Bag
Quantity 10 Ea.
Dimensions (Dia. x H) 100 x 15 mm
Width (Metric) 15 mm
Height (Metric) 100 mm
Diameter (Metric) 15 mm
Species Streptococcus pyogenes (group A)
Protein Source Yeast Extract
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.