Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ HardyCHROM™ MRSA Chromogenic media
SDP

Catalog No. 23001795 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Species:
Staphylococcus aureus
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
23-001-795 Staphylococcus aureus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-795 Supplier Hardy Diagnostics™ Supplier No. G307
Add to Cart
Add to Cart
This item is not returnable. View return policy

A selective and differential culture medium that facilitates the isolation and identification of methicillin resistant Staphylococcus aureus (MRSA) to aid in the prevention and control of MRSA infections in health care settings.

  • Test can be performed on anterior nares swabs from patients and healthcare workers to screen for MRSA colonization
  • This new chromogenic medium simplifies the identification of MRSA infections. MRSA strains grown in the presence of chromogenic substrates that produce deep pink to magenta colonies. Color development is bright, distinct, and easy to read
  • Highly sensitive and specific
  • Compatible with automation
  • Minimal training required

Specifications

Appearance Translucent
Color Light Amber
Content And Storage 2°C to 8°C
Additive Chromogenic Mixture, Inhibitory and Selective Agents
Detects Methicillin-resistant Staphylococcus aureus
Form Prepared Medium
Format Mono-Plate
Media Type Chromogenic Medium
Product Type Agar Plate
Target Organism Class Gram-positive Cocci
Test Type Microbial Identification
Fill Volume 10 mL
Formulation Sodium Chloride - 30.0gm, Peptone - 20.0gm, Chromogenic Mixture - 2.0gm, Inhibitory and Selective Agents - 2.5gm, Agar - 15.0gm
For Use With (Application) Isolation, Identification, and Qualitative Detection of MRSA Nasal Colonization
Incubation Conditions 35°C to 37°C
Incubation Time 24 to 48 hr.
Packaging Type Bag
Quantity 10 Ea.
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Staphylococcus aureus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.