Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ HardyCHROM™ O157 Chromogenic Medium
SDP

Catalog No. 23001744 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-744 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-744 Supplier Hardy Diagnostics™ Supplier No. G305
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

HardyCHROM™ O157 is a selective and differential medium recommended for the isolation of enterohemorrhagic E. coli O157 from food and environmental sources.

  • Chromogenic substances in the media facilitate detection by colony color
  • Not for human diagnostic use
  • HardyCHROM™ O157 provides an initial screen intended to isolate colonies for further testing
  • Confirm isolated purple-pink colonies with a latex aggulation test, antisera, or other test methods for complete identification
  • This media is to be stored and incubated in the dark

Specifications

Appearance Opaque
Color White to Off-white
Content And Storage Store in the Dark
Additive Chromogenic Mixture, Selective Agents
Detects Enterohemorrhagic E. coli O157
Form Prepared Medium
Format Plate
Media Type Chromogenic Media
Product Type Agar Plate
Target Organism Class E. coli
Test Type Microbial Identification
Fill Volume 18 mL
Formulation Peptones - 13.0gm, Chromogenic Mixture - 1.0gm, Selective Agents - 1.0gm, Agar - 15.0gm
For Use With (Application) Isolation of enterohemorrhagic E. coli O157
Incubation Conditions 35°C
Incubation Time 24 hr.
No. per Pack 10
Packaging Type Bag
Quantity 10 Plates
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Escherichia coli, Enterobacter aerogenes, Proteus mirabilis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.