Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ HardyCHROM™ SS NoPRO
SDP

Catalog No. 23001747 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-747 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-747 Supplier Hardy Diagnostics™ Supplier No. G327
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

HardyCHROM SS NoPro (no-Proteus) Agar is recommended for the selective isolation of Salmonella and Shigella spp. from stool. SS NoPro Agar is intended as a primary screening tool to distinguish Salmonella and Shigella spp. from non-pathogenic enteric bacteria based on colony color.

  • The enhanced inhibition of Proteus reduces the expense involved in working up non-pathogens that could mimic enteric pathogens
  • Further species confirmation of suspect colonies via conventional or automated methods is recommended
  • Less colony picking, subculturing, and identifications
  • No need for TSI, LIA, KIA tubes
  • Reduces the number of plates for primary stool setup

Specifications

Appearance Clear, Slightly Opalescent
Color Dark Pink
Content And Storage 2°C to 8°C
Additive Chromogenic Mixture, Selective Agents, pH Indicator
Detects Salmonella spp., Shigella spp.
Form Prepared Medium
Format Agar Plate
Media Type Chromogenic Agar
Product Type Agar Plate
Target Organism Class Gram-negative Bacteria
Test Type Microbial Differentiation
Fill Volume 18 mL
Formulation Carbohydrates - 20.0g, Peptones - 8.0g, Sodium Thiosulfate - 3.5g, Sodium Deoxycholate - 1.5g, Ferric Citrate - 1.0g, Chromogenic Mixture - 0.5g, Selective Agents - 0.15g, pH Indicator - 0.01g, Agar - 15.0g
For Use With (Application) Selective isolation and differentiation of Salmonella and Shigella spp.
Incubation Conditions 35°C
Incubation Time 24 hr.
No. per Pack 10
Packaging Type Bag
Quantity 10 Ea.
Dimensions (Dia. x H) 100 x 15 mm
Width (Metric) 100 mm
Height (Metric) 15 mm
Depth (Metric) 15 mm
Diameter (Metric) 100 mm
Species Salmonella enterica, Shigella sonnei, Escherichia coli
Protein Source Peptones
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.