Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ HardyCHROM™ Staphylococcus Aureus Chromogenic Medium
SDP

Catalog No. 23001750 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Appearance: Translucent
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-001-750 Appearance: Translucent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-750 Supplier Hardy Diagnostics™ Supplier No. G311
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

HardyCHROM™ Staph aureus is a chromogenic medium recommended for the isolation, differentiation, and enumeration of Staphylococcus aureus by colony color

  • HardyCHROM™ Staph aureus allows for the rapid and reliable detection of S. aureus from both clinical and food specimens within 24 hours
  • Selective agents inhibit the growth of gram-negative organisms, yeast, and some gram-positive cocci

Specifications

Appearance Translucent
Color Light Amber
Detects Staphylococcus aureus
Form Prepared Medium
Manufacturing Origin United States
Media Type Chromogenic Medium
Product Type Agar Plate
Target Organism Class Gram-positive Cocci
Test Type Microbial Detection
Fill Volume 18 mL
For Use With (Application) Isolation, Differentiation, Enumeration of Staphylococcus aureus
Incubation Conditions 35°C
Incubation Time 24 hr.
Packaging Type Box
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 100 mm
Diameter (Metric) 15 mm
Species Staphylococcus aureus, Staphylococcus species
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.