Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ HardyCHROM™ UTI Chromogenic Medium
SDP

Catalog No. 23001815 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Product Type: Agar Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-001-815 Product Type: Agar Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-815 Supplier Hardy Diagnostics™ Supplier No. G313
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

UTI is a differential culture medium that facilitates the isolation and differentiation of urinary tract pathogens, including gram-negative and gram-positive bacteria

  • The development of various colors, due to chromogenic substances in the media, allows for the differentiation of microorganisms from the primary set-up of a urine specimen
  • E. coli produces large magenta colonies (confirmatory, no further testing required)
  • Enterococcus faecalis/faecium produces small, turquoise-colored colonies
  • Pseudomonas spp. produce light yellow/green, translucent colonies
  • Klebsiella, Enterobacter, and Serratia spp. produce large, deep blue colonies
  • Staphylococcus saprophyticus produces opaque, pink colonies
  • Candida spp. produces small, white, moist colonies
  • Proteus, Morganella, and Providencia spp. produce clear to light yellow colonies with a diffuse golden-orange halo in the medium
  • Staphylococcus aureus produces opaque, white-colored colonies
  • Citrobacter spp. produce dark blue colonies often with a rose halo in the surrounding media

Specifications

Appearance Translucent
Color Light Off-white
Detects Urinary Tract Pathogens, Gram-negative Bacteria, Gram-positive Bacteria, Escherichia coli
Product Type Agar Plate
Target Organism Class Gram-positive Organisms, Gram-negative Organisms
Fill Volume 18 mL
For Use With (Application) Isolation and Differentiation of Urinary Tract Pathogens
Incubation Conditions 35°C
Incubation Time 24 hr.
No. per Pack 10
Quantity 10 Ea.
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Escherichia coli, Klebsiella pneumoniae, Enterococcus faecalis, Proteus species, Pseudomonas aeruginosa
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.