Learn More
Hardy Diagnostics™ HardyDisks™ Ceftazidime/Avibactam, CZA50, Avycaz
SDP
Catalog No. 23001801 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Quantity:
50 Pk.
Catalog No. Quantity
23-001-801 50 Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-801 Supplier Hardy Diagnostics™ Supplier No. Z9355
Add to Cart
Add to Cart
This item is not returnable. View return policy
HardyDisk AST's are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST) also known as disk diffusion or Kirby-Bauer testing.
  • HardyDisk™ AST Disks are used for semi-quantitative in vitro susceptibility testing by the agar diffusion test procedure (KirbyBauer) of rapidly growing and certain fastidious bacterial pathogens
  • HardyDisk™ AST Disks are prepared by impregnating high-quality 6mm diameter white filter paper disks with accurately determined amounts of antimicrobics or other chemotherapeutic agents
  • Disks are clearly marked on both sides with letters and numbers designating the agent and the drug content
  • Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing
Specifications
Product Type Antimicrobial Susceptibility Discs
Test Type Antibiotic Susceptibility Sensitivity
No. per Pack 50
Quantity 50 Pk.