Learn More
Hardy Diagnostics™ HardyDisks™ Ceftolozane/Tazobactam
SDP
Catalog No. 23001819 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
10
50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-819 10
23-001-793 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-001-819 Supplier Hardy Diagnostics™ Supplier No. Z9341
Add to Cart
Add to Cart
This item is not returnable. View return policy
HardyDisk AST's are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST) also known as disk diffusion or Kirby-Bauer testing.
  • HardyDisk™ AST Disks are used for semi-quantitative in vitro susceptibility testing by the agar diffusion test procedure (KirbyBauer) of rapidly growing and certain fastidious bacterial pathogens
  • HardyDisk™ AST Disks are prepared by impregnating high-quality 6mm diameter white filter paper disks with accurately determined amounts of antimicrobics or other chemotherapeutic agents
  • Disks are clearly marked on both sides with letters and numbers designating the agent and the drug content
Specifications
Product Type Antibiotic Assay Discs
Target Organism Class Fastidious
Test Type Antibiotic Susceptibility Sensitivity
No. per Pack 10