Learn More
Hardy Diagnostics™ HardyDisks™ Ceftolozane/Tazobactam
HardyDisk AST's are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST) also known as disk diffusion or Kirby-Bauer testing.
$16.17 - $58.30
Specifications
Product Type | Antibiotic Assay Discs |
---|---|
Target Organism Class | Fastidious |
Test Type | Antibiotic Susceptibility Sensitivity |
Catalog Number | Mfr. No. | No. per Pack | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. per Pack | Price | Quantity & Availability | |||||
23-001-819
![]() |
Hardy Diagnostics™
Z9341 |
10 |
Pack of 50 for $16.17
|
|
|||||
23-001-793
![]() |
Hardy Diagnostics™
Z9345 |
50 |
Pack of 250 for $58.30
|
|
|||||
Description
- HardyDisk™ AST Disks are used for semi-quantitative in vitro susceptibility testing by the agar diffusion test procedure (KirbyBauer) of rapidly growing and certain fastidious bacterial pathogens
- HardyDisk™ AST Disks are prepared by impregnating high-quality 6mm diameter white filter paper disks with accurately determined amounts of antimicrobics or other chemotherapeutic agents
- Disks are clearly marked on both sides with letters and numbers designating the agent and the drug content
Specifications
Antibiotic Assay Discs | |
Fastidious | |
Antibiotic Susceptibility Sensitivity |