Learn More
Hardy Diagnostics™ HardyFlask™ Emmon's Sabouraud Dextrose Agar (SabDex)
SDP
Catalog No. 23001827 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Quantity:
10 mL
Catalog No. Quantity
23-001-827 10 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-001-827 Supplier Hardy Diagnostics™ Supplier No. X57
Add to Cart
Add to Cart
This item is not returnable. View return policy
Emmon's Sabdex Agar is recommended for the isolation, cultivation, and maintenance of non-pathogenic and pathogenic species of fungi and yeasts.
  • Lightweight - For reduced shipping cost and less chance of breakage.
  • Stackable - For increased conservation of space, both in incubator and refrigerator. No special storage racks are needed.
  • Interlocking - For increased stability when incubating and transporting.
  • Large Surface Area - For the inoculum. Almost double the area! better isolation of colonies.
  • Slanted Neck - Facilitates easy inoculation of the flat media surface.
  • Improved Clarity - For better visibility of the growing culture, both front and back.
Specifications
Manufacturing Origin United States
Media Type Sabouraud Dextrose Agar
Product Type Agar Plate
Sterility Non-sterile
Target Organism Class Fungi, Yeast
Test Type Microbial Detection
Quantity 10 mL