Learn More
Cayman Chemical H-Arg-Gly-Asp-Cys-OH (trifluoroacetate salt), Purity: ≥95%, 100 mg
SDP
Catalog No. 502946313
Encompass_Preferred
An RGD-containing tetrapeptide; inhibits the binding of fibrinogen to endothelial cells and ADP-stimulated platelets (IC50s = 320 and 35 µM, respectively); implantation of titanium rods coated with H-Arg-Gly-Asp-Cys-OH increases bone formation in rat femurs; has been conjugated to polyethylenimine to improve gene transfection efficiency
Catalog No. 50-294-6313 Supplier Cayman Chemical Supplier No. 34543100 MG
An RGD-containing tetrapeptide; inhibits the binding of fibrinogen to endothelial cells and ADP-stimulated platelets (IC50s = 320 and 35 µM, respectively); implantation of titanium rods coated with H-Arg-Gly-Asp-Cys-OH increases bone formation in rat femurs; has been conjugated to polyethylenimine to improve gene transfection efficiency