Learn More
MilliporeSigma™ Harleco™ Histology/Papanicolaou Stains: EA-50 Papanicolaou Staining Solution
Catalog No. 23750069 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4000mL
Catalog No. Quantity
23-750069 4000mL
1 options
This item is not returnable. View return policy
Catalog No. 23-750069 Supplier MilliporeSigma™ Supplier No. 7062X85
This item is not returnable. View return policy
Harleco™ Histology/Papanicolaou Stains
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Cytoplasmic counterstain for pap staining technique
Quantity 4000mL
Product Type Papanicolaou Stain