Learn More
NEOGEN Harlequin Tryptone Bile Glucuronide Agar (TBX), 500 g
SDP
Catalog No. 502015954
Encompass_Preferred
Harlequin Tryptone Bile Glucuronide Agar (TBX) is used for the enumeration of E. coli in food and animal feed and is not intended for use in the diagnosis of disease or other conditions in humans. Tryptone Bile Glucuronide Agar is recommended by ISO 16649-1:2001, ISO 16649-2:2001 and ISO 16649-3:2015 for enumeration of β-glucuronidase positive Escherichia coli in products intended for human consumption or for the feeding of animals. The medium contains tryptone as a source of nitrogen and amino acids, bile salts act as a selective against Gram-positive organisms and agar is the gelling agent. 5-bromo-4-chloro-3-indolyl-β-D-glucuronide (X-glucuronide) is a chromogen for the detection of the β-D-glucuronidase enzyme which is present in most E. coli* but not other coliforms.*96-97% of E.coli are β-D-glucuronidase positive. A notable exception is E. coli O157:H7  For more information visit https://www.neogen.com/solutions/Microbiology/harlequin-tryptone-bile-glucuronide-agar-tbx
Catalog No. 50-201-5954 Supplier NEOGEN Supplier No. 700004780
Harlequin Tryptone Bile Glucuronide Agar (TBX) is used for the enumeration of E. coli in food and animal feed and is not intended for use in the diagnosis of disease or other conditions in humans. Tryptone Bile Glucuronide Agar is recommended by ISO 16649-1:2001, ISO 16649-2:2001 and ISO 16649-3:2015 for enumeration of β-glucuronidase positive Escherichia coli in products intended for human consumption or for the feeding of animals. The medium contains tryptone as a source of nitrogen and amino acids, bile salts act as a selective against Gram-positive organisms and agar is the gelling agent. 5-bromo-4-chloro-3-indolyl-β-D-glucuronide (X-glucuronide) is a chromogen for the detection of the β-D-glucuronidase enzyme which is present in most E. coli* but not other coliforms.*96-97% of E.coli are β-D-glucuronidase positive. A notable exception is E. coli O157:H7  For more information visit https://www.neogen.com/solutions/Microbiology/harlequin-tryptone-bile-glucuronide-agar-tbx