Learn More
Medchemexpress LLC Harmalol hydrochloride | 6028-07-5 | 1 ML
SDP
Catalog No. 5000420259
Encompass_Preferred
Harmalol hydrochloride is a beta carboline alkaloid found in medicinal plants like *Peganum harmala*. It is a main metabolite of Harmaline and significantly inhibits the dioxin-mediated induction of CYP1A1 at both transcriptional and posttranslational levels. This compound also exhibits antioxidant and hydroxyl radical-scavenging properties.
  • Purity: 99.91%
  • Inhibits dioxin-mediated induction of CYP1A1
  • Possesses antioxidant and hydroxyl radical-scavenging properties
  • For research use only
Catalog No. 50-004-20259 Supplier Medchemexpress LLC Supplier No. HYN2625A10MM/1ML
Harmalol hydrochloride is a beta carboline alkaloid found in medicinal plants like *Peganum harmala*. It is a main metabolite of Harmaline and significantly inhibits the dioxin-mediated induction of CYP1A1 at both transcriptional and posttranslational levels. This compound also exhibits antioxidant and hydroxyl radical-scavenging properties.
  • Purity: 99.91%
  • Inhibits dioxin-mediated induction of CYP1A1
  • Possesses antioxidant and hydroxyl radical-scavenging properties
  • For research use only