Learn More
Medchemexpress LLC Harmalol hydrochloride | 6028-07-5 | 100 MG
SDP
Catalog No. 5000421667
Encompass_Preferred
Harmalol hydrochloride is a beta carboline alkaloid primarily found in medicinal plants such as *Peganum harmala*. This compound serves as the main metabolite of Harmaline and has been shown to significantly inhibit the dioxin-mediated induction of CYP1A1 at both transcriptional and posttranslational levels. Furthermore, it exhibits notable antioxidant and hydroxyl radical-scavenging properties, making it of interest for various research applications.
  • Main metabolite of Harmaline
  • Inhibits dioxin-mediated induction of CYP1A1
  • Exhibits antioxidant and hydroxyl radical-scavenging properties
  • Suitable for research applications
Catalog No. 50-004-21667 Supplier Medchemexpress LLC Supplier No. HYN2625A100MG
Harmalol hydrochloride is a beta carboline alkaloid primarily found in medicinal plants such as *Peganum harmala*. This compound serves as the main metabolite of Harmaline and has been shown to significantly inhibit the dioxin-mediated induction of CYP1A1 at both transcriptional and posttranslational levels. Furthermore, it exhibits notable antioxidant and hydroxyl radical-scavenging properties, making it of interest for various research applications.
  • Main metabolite of Harmaline
  • Inhibits dioxin-mediated induction of CYP1A1
  • Exhibits antioxidant and hydroxyl radical-scavenging properties
  • Suitable for research applications