Learn More
Medchemexpress LLC Harmalol hydrochloride | 6028-07-5 | 25 MG
SDP
Catalog No. 5000421773
Encompass_Preferred
Harmalol hydrochloride is a beta carboline alkaloid found in medicinal plants like *Peganum harmala*. It is the main metabolite of Harmaline and significantly inhibits the dioxin-mediated induction of CYP1A1 at both transcriptional and posttranslational levels. This compound also exhibits antioxidant and hydroxyl radical-scavenging properties and is intended for research use only.
  • Beta carboline alkaloid
  • Found in medicinal plants such as *Peganum harmala*
  • Main metabolite of Harmaline
  • Inhibits CYP1A1 induction
  • Exhibits antioxidant properties
  • Possesses hydroxyl radical-scavenging properties
  • For research use only
Catalog No. 50-004-21773 Supplier Medchemexpress LLC Supplier No. 25MGHYN2625A
Harmalol hydrochloride is a beta carboline alkaloid found in medicinal plants like *Peganum harmala*. It is the main metabolite of Harmaline and significantly inhibits the dioxin-mediated induction of CYP1A1 at both transcriptional and posttranslational levels. This compound also exhibits antioxidant and hydroxyl radical-scavenging properties and is intended for research use only.
  • Beta carboline alkaloid
  • Found in medicinal plants such as *Peganum harmala*
  • Main metabolite of Harmaline
  • Inhibits CYP1A1 induction
  • Exhibits antioxidant properties
  • Possesses hydroxyl radical-scavenging properties
  • For research use only