Learn More
Medchemexpress LLC Harmalol hydrochloride | 6028-07-5 | 5 MG
SDP
Catalog No. 5000421051
Encompass_Preferred
Harmalol hydrochloride is a beta carboline alkaloid found in medicinal plants like *Peganum harmala*. It is the main metabolite of Harmaline and significantly inhibits the dioxin-mediated induction of CYP1A1 at both transcriptional and posttranslational levels. This compound also exhibits antioxidant and hydroxyl radical-scavenging properties.
  • Main metabolite of Harmaline.
  • Significantly inhibits dioxin-mediated induction of CYP1A1 at transcriptional and posttranslational levels.
  • Possesses antioxidant properties.
  • Exhibits hydroxyl radical-scavenging properties.
Catalog No. 50-004-21051 Supplier Medchemexpress LLC Supplier No. HYN2625A5MG
Harmalol hydrochloride is a beta carboline alkaloid found in medicinal plants like *Peganum harmala*. It is the main metabolite of Harmaline and significantly inhibits the dioxin-mediated induction of CYP1A1 at both transcriptional and posttranslational levels. This compound also exhibits antioxidant and hydroxyl radical-scavenging properties.
  • Main metabolite of Harmaline.
  • Significantly inhibits dioxin-mediated induction of CYP1A1 at transcriptional and posttranslational levels.
  • Possesses antioxidant properties.
  • Exhibits hydroxyl radical-scavenging properties.