Learn More
Medchemexpress LLC Harmalol hydrochloride | 6028-07-5 | 50 MG
SDP
Catalog No. 5000421423
Encompass_Preferred
Harmalol hydrochloride is a beta carboline alkaloid found in medicinal plants such as Peganum harmala, and is the main metabolite of Harmaline. This compound inhibits the dioxin-mediated induction of CYP1A1 at both transcriptional and posttranslational levels, and demonstrates antioxidant and hydroxyl radical-scavenging properties. It is intended for research use only.
  • Found in medicinal plants, including Peganum harmala
  • Main metabolite of Harmaline
  • Inhibits dioxin-mediated induction of CYP1A1
  • Exhibits antioxidant properties
  • Demonstrates hydroxyl radical-scavenging properties
  • For research use only
  • Solid appearance, light yellow to yellow color
Catalog No. 50-004-21423 Supplier Medchemexpress LLC Supplier No. HYN2625A50MG
Harmalol hydrochloride is a beta carboline alkaloid found in medicinal plants such as Peganum harmala, and is the main metabolite of Harmaline. This compound inhibits the dioxin-mediated induction of CYP1A1 at both transcriptional and posttranslational levels, and demonstrates antioxidant and hydroxyl radical-scavenging properties. It is intended for research use only.
  • Found in medicinal plants, including Peganum harmala
  • Main metabolite of Harmaline
  • Inhibits dioxin-mediated induction of CYP1A1
  • Exhibits antioxidant properties
  • Demonstrates hydroxyl radical-scavenging properties
  • For research use only
  • Solid appearance, light yellow to yellow color