Learn More
Medchemexpress LLC Harmalol hydrochloride | 6028-07-5 | 99.9% | 236.70 g/mol | C12H13ClN2O | 10 MG
SDP
Catalog No. 5000289664
Encompass_Preferred
Harmalol hydrochloride is the hydrochloride salt of harmalol, a beta-carboline alkaloid supplied for research use. It is reported to inhibit dioxin-mediated induction of CYP1A1 and to exhibit antioxidant and hydroxyl radical-scavenging activity.
  • High purity: 99.91%.
  • Hydrochloride salt form, solid, suitable for biochemical and pharmacological studies.
  • Molecular formula C12H13ClN2O; molecular weight 236.70 g/mol.
  • Provided in small research quantities appropriate for assay development and analytical testing.
  • Store sealed at 4°C, protected from moisture and light; in solution store at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-89664 Supplier Medchemexpress LLC Supplier No. HYN2625A10MG
Harmalol hydrochloride is the hydrochloride salt of harmalol, a beta-carboline alkaloid supplied for research use. It is reported to inhibit dioxin-mediated induction of CYP1A1 and to exhibit antioxidant and hydroxyl radical-scavenging activity.
  • High purity: 99.91%.
  • Hydrochloride salt form, solid, suitable for biochemical and pharmacological studies.
  • Molecular formula C12H13ClN2O; molecular weight 236.70 g/mol.
  • Provided in small research quantities appropriate for assay development and analytical testing.
  • Store sealed at 4°C, protected from moisture and light; in solution store at -80°C (6 months) or -20°C (1 month).