Learn More
Medchemexpress LLC Harmane | 486-84-0 | MFCD00004957 | 182.23 g/mol | C12H10N2 | 250 MG
SDP
Catalog No. 5000213180
Encompass_Preferred
Harmane is a beta-carboline alkaloid research compound with reported activity at imidazoline, adrenergic, monoamine oxidase, nicotinic acetylcholine, GABA, and opioid receptors. Supplied as a powder for biochemical and pharmacological studies, it is readily soluble in DMSO and accompanied by storage and solubility data for experimental use.
  • Beta-carboline alkaloid research compound.
  • Documented activity across multiple neurotransmitter targets.
  • High solubility in DMSO (100 mg/mL) for in vitro work.
  • Storage guidelines for powder and solutions to preserve stability.
  • Chemical identifiers and physicochemical properties provided.
Catalog No. 50-002-13180 Supplier Medchemexpress LLC Supplier No. HY101392250MG
Harmane is a beta-carboline alkaloid research compound with reported activity at imidazoline, adrenergic, monoamine oxidase, nicotinic acetylcholine, GABA, and opioid receptors. Supplied as a powder for biochemical and pharmacological studies, it is readily soluble in DMSO and accompanied by storage and solubility data for experimental use.
  • Beta-carboline alkaloid research compound.
  • Documented activity across multiple neurotransmitter targets.
  • High solubility in DMSO (100 mg/mL) for in vitro work.
  • Storage guidelines for powder and solutions to preserve stability.
  • Chemical identifiers and physicochemical properties provided.