Learn More
Medchemexpress LLC Harmane-d1 | 486-84-0 | 98.0% | 183.23 | C12H9DN2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY101392S10MG
Encompass_Preferred
Harmane-d1 is the deuterium-labeled isotopic standard of harmane supplied as a solid for laboratory research. It is intended for use as an analytical reference or tracer in studies involving imidazoline, adrenergic, and monoamine oxidase biology, and is provided in milligram-scale packaging for analytical applications.
  • Deuterium-labeled isotopic standard for harmane.
  • High purity (98.0%) suitable for analytical use.
  • Molecular weight 183.23 g/mol and formula C12H9DN2.
  • Provided as a solid in a 10 mg quantity.
  • Documentation includes data sheet and safety data sheet.
Catalog No. 50-002-11828