Learn More
Medchemexpress LLC Harmane hydrochloride | 21655-84-5 | 99.4% | 218.68 g/mol | C12H11ClN2 | 5 MG
SDP
Catalog No. 5000213217
Encompass_Preferred
Harmane hydrochloride is the hydrochloride salt of a β-carboline alkaloid used in neuropharmacology research. It modulates imidazoline receptors and monoamine oxidase and is supplied as a high-purity analytical reagent for assays, receptor profiling, and mechanistic studies.
  • High purity (99.35%) suitable for analytical use.
  • Molecular formula C12H11ClN2 and molecular weight 218.68 g/mol.
  • Provided in small vials for assay and analytical workflows.
  • Active compound for studies of imidazoline receptors and MAO-related pathways.
  • Certificate of analysis and safety data sheet available for quality and safety information.
Catalog No. 50-002-13217 Supplier Medchemexpress LLC Supplier No. HY101392A5MG
Harmane hydrochloride is the hydrochloride salt of a β-carboline alkaloid used in neuropharmacology research. It modulates imidazoline receptors and monoamine oxidase and is supplied as a high-purity analytical reagent for assays, receptor profiling, and mechanistic studies.
  • High purity (99.35%) suitable for analytical use.
  • Molecular formula C12H11ClN2 and molecular weight 218.68 g/mol.
  • Provided in small vials for assay and analytical workflows.
  • Active compound for studies of imidazoline receptors and MAO-related pathways.
  • Certificate of analysis and safety data sheet available for quality and safety information.