Learn More
Medchemexpress LLC HY-N0737A 500mg Medchemexpress, Harmine CAS:442-51-3 Purity:>98%
SDP
Catalog No. 502029418
Encompass_Preferred
Medchemexpress, HY-N0737A 500mg Harmine CAS:442-51-3 Harmine is a natural dual-specificity tyrosine phosphorylation-regulated kinase (DYRK) inhibitor with anticancer and anti-inflammatory activities. Harmine has a high affinity of 5-HT2A serotonin receptor, with an Ki of 397 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9418 Supplier Medchemexpress LLC Supplier No. HYN0737A500MG
Medchemexpress, HY-N0737A 500mg Harmine CAS:442-51-3 Harmine is a natural dual-specificity tyrosine phosphorylation-regulated kinase (DYRK) inhibitor with anticancer and anti-inflammatory activities. Harmine has a high affinity of 5-HT2A serotonin receptor, with an Ki of 397 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.